accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107-37-9

IUPAC Name: trichloro(prop-2-en-1-yl)silane
Synonyms: allytrichlorosilane
Molecular Weight (g/mol): 175.51
Molecular Formula: C3H5Cl3Si
InChi Key: HKFSBKQQYCMCKO-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)CC=C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)