Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyltrichlorosilane, 95%

Catalog No. AAL0370314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0370314 25 g
AAL0370306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0370314 Supplier Thermo Scientific Chemicals Supplier No. L0370314
  • $221.00
    $176.65 / Each of 1

    Save $44.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 107-37-9 | C3H5Cl3Si | 175.508 g/mol

Allyltrichlorosilane acts as a reagent that produces homoallylic alcohols upon reaction with aldehydes in DMF. It is used to make silicones and glass fiber fininshes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyltrichlorosilane acts as a reagent that produces homoallylic alcohols upon reaction with aldehydes in DMF. It is used to make silicones and glass fiber fininshes.

Solubility
Soluble in THF, toluene, benzene. Reacts with water.

Notes
Moisture Sensitive. Store at 4°C. Store away from oxidizing agents and moisture. Incompatible with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-37-9
Molecular Formula C3H5Cl3Si
Molecular Weight (g/mol) 175.508
MDL Number MFCD00000486
InChI Key HKFSBKQQYCMCKO-UHFFFAOYSA-N
Synonym allyltrichlorosilane, silane, trichloro-2-propenyl, silane, allyltrichloro, silane, trichloroallyl, allyl trichlorosilane, propen-3-yltrichlorosilane, allytrichlorosilane, allylsilicone trichloride, unii-ub3n98803n, silane, trichloro-2-propen-1-yl
PubChem CID 7867
IUPAC Name trichloro(prop-2-enyl)silane
SMILES C=CC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 117°C to 118°C
Quantity 25 g
Flash Point 18°C (64°F)
Refractive Index 1.449
Sensitivity Moisture sensitive
Density 1.211
Linear Formula CH2=CHCH2SiCl3
UN Number UN1724
Beilstein 1743449
Solubility Information Soluble in THF,toluene,benzene. Reacts with water.
Formula Weight 175.52
Percent Purity 95%
Chemical Name or Material Allyltrichlorosilane
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 203-485-9
RTECSNumber VV1530000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.