accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3COSiCH3H3COSiCH3CH3H3C

CAS RN 107-51-7

IUPAC Name: 2,2,4,4,6,6-hexamethyl-3,5-dioxa-2,4,6-trisilaheptane
Synonyms: octamethyltrisiloxane
Molecular Weight (g/mol): 236.53
Molecular Formula: C8H24O2Si3
InChi Key: CXQXSVUQTKDNFP-UHFFFAOYSA-N
SMILES: C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)