Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Octamethyltrisiloxane

Catalog No. 2357095ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-570-95ML 5 mL
23-570-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-570-95ML Supplier Sigma Aldrich Supplier No. 2357095ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Octamethyltrisiloxane

Specifications

CAS 107-51-7
Melting Point -82°C
Density 0.82 g/mL at 25°C
Boiling Point 153°C
Molecular Formula C8H24O2Si3
Linear Formula [(CH3)3SiO]2Si(CH3)2
Refractive Index n20/D 1.384
MDL Number MFCD00008264
Quantity 5 mL
Molecular Weight (g/mol) 236.53 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.