CAS RN 107-54-0
CAS RN 107-54-0
IUPAC Name:
3,5-dimethylhex-1-yn-3-ol
Synonyms:
3,5-dimethylhex-1-yn-3-ol
Molecular Weight (g/mol):
126.2
Molecular Formula:
C8H14O
InChi Key:
NECRQCBKTGZNMH-UHFFFAOYNA-N
SMILES:
CC(C)CC(C)(O)C#C
1
–
1
of
1
results
1
3,5-Dimethyl-1-hexyn-3-ol 98.0+%, TCI America™
CAS: 107-54-0 Molecular Formula: C8H14O Molecular Weight (g/mol): 126.20 MDL Number: MFCD00008941 InChI Key: NECRQCBKTGZNMH-UHFFFAOYNA-N Synonym: 3,5-dimethyl-1-hexyn-3-ol,surfynol,surfynol 61,1-hexyn-3-ol, 3,5-dimethyl,surfynol van,sialyl-3-paragloboside,acmc-2098wy,dsstox_cid_21472,dsstox_rid_79746,dsstox_gsid_41472 PubChem CID: 61018 IUPAC Name: 3,5-dimethylhex-1-yn-3-ol SMILES: CC(C)CC(C)(O)C#C