Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,5-Dimethyl-1-hexyn-3-ol

Catalog No. 278394500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-839-4500ML 500 mL
27-839-4100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 27-839-4500ML Supplier Sigma Aldrich Supplier No. 278394500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Bimolecular rate constant for the reaction of the hydroxyl radical (oh) with 3,5-dimethyl-1-hexyn-3-ol has been been measured using the relative rate technique. reaction of co2 with 3,5-dimethyl-1-hexyn-3-ol catalyzed by cucl in different ionic liquids and solvents has been investigated.3,5-dimethyl-1-hexyn-3-ol may be used to synthesize:
  • 3,5-dimethyl-1-phenyl-1-hexen-3-ol via one-pot palladium-mediated hydrostannylation/stille cross-coupling
  • 3,5-dimethyl-1-hexyn-3-acetate via esterification with acetic anhydride in a neutral ionic liquid (1-butyl-3-methylimidazolium tetrafluoroborate)
  • 3,5-dimethyl-3-hydroxy-1-hexen-1-yl benzoate via anti-markovnikov addition of benzoic acid

Specifications

CAS 107-54-0
Density 0.859 g/mL (at 25°C (literature))
Boiling Point 150°C to 151°C (lit.)
Molecular Formula C8H14O
Refractive Index n20/D 1.434 (literature)
Linear Formula (CH3)2 CHCH2C(CH3)(OH)C///CH
MDL Number MFCD00008941
Quantity 500 mL
Molecular Weight (g/mol) 126.2
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.