accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107-84-6

IUPAC Name: 1-chloro-3-methylbutane
Synonyms: butane, 1-chloro-3-methyl-
Molecular Weight (g/mol): 106.59
Molecular Formula: C5H11Cl
InChi Key: CZHLPWNZCJEPJB-UHFFFAOYSA-N
SMILES: CC(C)CCCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results