Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-3-methylbutane 98.0+%, TCI America™
SDP

Catalog No. C0199500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0199500ML 500 mL
C019925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0199500ML Supplier TCI America Supplier No. C0199500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 107-84-6
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.593
MDL Number MFCD00013693
InChI Key CZHLPWNZCJEPJB-UHFFFAOYSA-N
Synonym isoamyl chloride, butane, 1-chloro-3-methyl, 3-methylbutyl chloride, isopentyl chloride, 4-chloro-2-methylbutane, 1-chloro-3,3-dimethylpropane, unii-87d3zl9f7a, isoamylchlorid, acmc-2098xt, 1-chloro-3-methyl-butane
PubChem CID 7893
IUPAC Name 1-chloro-3-methylbutane
SMILES CC(C)CCCl

Specifications

Color Colorless
Boiling Point 100°C
Quantity 500 mL
UN Number 1107
Formula Weight 106.59
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-3-methylbutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.