accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107-85-7

IUPAC Name: 3-methylbutan-1-amine
Synonyms: isoamylamine leucamine isobutylcarbylamine 1-aminoisopentane
Molecular Weight (g/mol): 87.17
Molecular Formula: C5H13N
InChi Key: BMFVGAAISNGQNM-UHFFFAOYSA-N
SMILES: CC(C)CCN
Boiling Point 
  • (1)
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (4)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)