Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isopentylamine

Catalog No. 12681010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-681-010G 10 g
12-681-0100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-681-010G Supplier Sigma Aldrich Supplier No. 12681010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Isopentylamine is produced by proteus mirabilis
  • It is present in kola nut varieties of nigerian origin

Specifications

CAS 107-85-7
Density 0.751 g/mL (at 25°C (literature))
Boiling Point 95°C to 97°C (lit.)
Molecular Formula C5H13N
Linear Formula (CH3)2 CHCH2CH2NH2
Refractive Index n20/D 1.408 (literature)
MDL Number MFCD00008203
Quantity 10 g
Synonym 1-Amino-3-methylbutane; Isoamylamine
Molecular Weight (g/mol) 87.16
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.