accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHO(E)

CAS RN 107-93-7

IUPAC Name: (2E)-but-2-enoic acid
Synonyms: butenoic acid BEO kyselina krotonova α-crotonic acid solid crotonic acid
Molecular Weight (g/mol): 86.09
Molecular Formula: C4H6O2
InChi Key: LDHQCZJRKDOVOX-NSCUHMNNSA-N
SMILES: C\C=C\C(O)=O
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)