accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHO

CAS RN 1070-34-4

IUPAC Name: 4-ethoxy-4-oxobutanoic acid
Synonyms: butanedioic acid, monoethyl ester
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: LOLKAJARZKDJTD-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC(O)=O
Boiling Point 
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)