Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mono-Ethyl succinate, 95%

Catalog No. AC436880050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436880050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC436880050 Supplier Thermo Scientific Chemicals Supplier No. 436880050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1070-34-4 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-34-4
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
InChI Key LOLKAJARZKDJTD-UHFFFAOYSA-N
Synonym monoethyl succinate, ethyl hydrogen succinate, mono-ethyl succinate, butanedioic acid, monoethyl ester, butanedioic acid, 1-ethyl ester, succinic acid monoethyl ester, monoethyl butanedioate, bernsteinsaureathylester, succinic acid monoethyl, succinic acid ethyl ester
PubChem CID 70610
IUPAC Name 4-ethoxy-4-oxobutanoic acid
SMILES CCOC(=O)CCC(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 8°C
Color Colorless
Density 1.1410g/mL
Elemental Analysis 46.4 to 52.3% C, 41.2 to 46.4% O
Boiling Point 248°C
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.432
Quantity 5 g
Specific Gravity 1.141
Formula Weight 146.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material mono-Ethyl succinate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 213-973-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.