accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHNPOHOHO

CAS RN 1071-83-6

IUPAC Name: 2-[(phosphonomethyl)amino]acetic acid
Synonyms: glyphosate epa pesticide chemical code 417300 glyphosate, dilithium salt magnesium salt (2:1) glyphosate glyphosate, zinc salt roundup max calcium salt glyphosate magnesium salt glyphosate monosodium salt glyphosate glyphosate, monopotassium salt
Molecular Weight (g/mol): 169.07
Molecular Formula: C3H8NO5P
InChi Key: XDDAORKBJWWYJS-UHFFFAOYSA-N
SMILES: OC(=O)CNCP(O)(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Solvent or Matrix 
  • (1)
  • (2)
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (6)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)