Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyphosate, >95% min., Ultra Scientific

Catalog No. USPST1850
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PST-1850 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PST-1850 Supplier Agilent Technologies Supplier No. PST1850
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1071-83-6
Molecular Formula C3H8NO5P
Molecular Weight (g/mol) 169.07
MDL Number MFCD00055350
InChI Key XDDAORKBJWWYJS-UHFFFAOYSA-N
PubChem CID 3496
ChEBI CHEBI:27744
IUPAC Name 2-[(phosphonomethyl)amino]acetic acid
SMILES OC(=O)CNCP(O)(O)=O

Specifications

CAS Max % 100.0
Assay Percent Range >95%
Quantity 100 mg
Chemical Name or Material Glyphosate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.