accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NHNOONHNH2

CAS RN 1071-93-8

IUPAC Name: hexanedihydrazide
Synonyms: adipohydrazide
Molecular Weight (g/mol): 174.2
Molecular Formula: C6H14N4O2
InChi Key: IBVAQQYNSHJXBV-UHFFFAOYSA-N
SMILES: NNC(=O)CCCCC(=O)NN
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)