Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adipylhydrazide, MP Biomedicals

Catalog No. ICN213452 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213452 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213452 Supplier MP Biomedicals, Inc Supplier No. 0521345210

Chemical Identifiers

CAS 1071-93-8
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00007614
InChI Key IBVAQQYNSHJXBV-UHFFFAOYSA-N
Synonym adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m
PubChem CID 66117
IUPAC Name hexanedihydrazide
SMILES NNC(=O)CCCCC(=O)NN

Specifications

Quantity 10 g
Formula Weight 174.2
Chemical Name or Material Adipyl Hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.