CAS RN 107135-55-7
IUPAC Name:
2-bromo-3-iodo-1,4-dimethoxybenzene
Synonyms:
2-bromo-3-iodo-1,4-dimethoxybenzene
Molecular Weight (g/mol):
342.96
Molecular Formula:
C8H8BrIO2
InChi Key:
RYNYTPCCJBVMOP-UHFFFAOYSA-N
SMILES:
COC1=C(Br)C(I)=C(OC)C=C1
1
–
1
of
1
results
1
2-Bromo-3-iodo-1,4-dimethoxybenzene, 97%, Thermo Scientific™
CAS: 107135-55-7 Molecular Formula: C8H8BrIO2 Molecular Weight (g/mol): 342.96 MDL Number: MFCD22380710 InChI Key: RYNYTPCCJBVMOP-UHFFFAOYSA-N Synonym: 1-bromo-2-iodo-3,6-dimethoxybenzene,1-bromo-3,6-dimethoxy-2-iodobenzene PubChem CID: 98001135 IUPAC Name: 2-bromo-3-iodo-1,4-dimethoxybenzene SMILES: COC1=C(Br)C(I)=C(OC)C=C1