accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1072-43-1

CH3S

CAS RN 1072-43-1

IUPAC Name: 2-methylthiirane
Synonyms: thiirane, methyl-
Molecular Weight (g/mol): 74.14
Molecular Formula: C3H6S
InChi Key: MBNVSWHUJDDZRH-UHFFFAOYNA-N
SMILES: CC1CS1
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)