Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene sulfide, Thermo Scientific™

Catalog No. AC131570250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC131570250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC131570250 Supplier Thermo Scientific Chemicals Supplier No. 131570250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-43-1
Molecular Formula C3H6S
Molecular Weight (g/mol) 74.14
MDL Number MFCD00005160
InChI Key MBNVSWHUJDDZRH-UHFFFAOYNA-N
Synonym propylene sulfide, thiirane, methyl, thiirane, 2-methyl, propene sulfide, 1,2-epithiopropane, propylene sulphide, propylene episulfide, 2-methylthiacyclopropane, propane, 1,2-epithio, methylthiirane
PubChem CID 14072
IUPAC Name 2-methylthiirane
SMILES CC1CS1

Specifications

Density 0.94g/mL
Assay Percent Range 98%
Quantity 25 g
Beilstein 17, II, 15
Formula Weight 74.14
Chemical Name or Material Propylene sulfide, 98%
Hazard Category Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.