accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1072-47-5

IUPAC Name: 4-methyl-1,3-dioxolane
Synonyms: 4-methyl-1,3-dioxolane
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: SBUOHGKIOVRDKY-UHFFFAOYNA-N
SMILES: CC1COCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results