accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHN

CAS RN 1072-62-4

IUPAC Name: 2-ethyl-1H-imidazole
Synonyms: 2-ethylimidazole
Molecular Weight (g/mol): 96.13
Molecular Formula: C5H8N2
InChi Key: PQAMFDRRWURCFQ-UHFFFAOYSA-N
SMILES: CCC1=NC=CN1
Percent Purity 
  • (2)
  • (4)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (2)
  • (2)
  • (4)
Physical Form 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (6)
  • (2)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)