Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylimidazole, 99%, Thermo Scientific™

Catalog No. AC219280000
Catalog No. AC219280000 Supplier Thermo Scientific Chemicals Supplier No. 219280000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-62-4
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005192
InChI Key PQAMFDRRWURCFQ-UHFFFAOYSA-N
Synonym 2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972
PubChem CID 66130
IUPAC Name 2-ethyl-1H-imidazole
SMILES CCC1=NC=CN1

Specifications

Melting Point 83.0°C to 87.0°C
Boiling Point 268.0°C
Flash Point 159°C
Solubility Information Solubility in water: 617g/L (20°C). Other solubilities: soluble in many organic solvents
Formula Weight 96.13
Chemical Name or Material 2-Ethylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.