accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOHOOHHOHNOOHOHOOHOHOHOH

CAS RN 1072896-38-8

IUPAC Name: 4-hydroxy-5-(2-hydroxyacetamido)-2-{[3,4,5-trihydroxy-6-(4-methoxyphenoxy)oxan-2-yl]methoxy}-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Synonyms: 4-hydroxy-5-(2-hydroxyacetamido)-2-{[3,4,5-trihydroxy-6-(4-methoxyphenoxy)oxan-2-yl]methoxy}-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Molecular Weight (g/mol): 593.54
Molecular Formula: C24H35NO16
InChi Key: QDQXPAOHAVZJRT-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COC3(CC(O)C(NC(=O)CO)C(O3)C(O)C(O)CO)C(O)=O)C(O)C(O)C2O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results