Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neu5Gc alpha(2-6)Gal beta MP Glycoside 97.0+%, TCI America™
SDP

Catalog No. N07945MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07945MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N07945MG Supplier TCI America Supplier No. N07945MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1072896-38-8
Molecular Formula C24H35NO16
Molecular Weight (g/mol) 593.54
MDL Number MFCD10566924
InChI Key QDQXPAOHAVZJRT-UHFFFAOYNA-N
Synonym 4-Methoxyphenyl 6-O-[N-(Hydroxyacetyl)-alpha-neuraminosyl]-beta-D-galactopyranoside, 4-Methoxyphenyl O-(5-Hydoxyacetamido-3,5-dideoxy-D-glycero-alpha-D-galacto-2-nonulopyranosonic Acid)-(2-6)-beta-D-galactopyranoside, Neu5Gc alpha(2-6)Gal-beta-MP
PubChem CID 133554174
IUPAC Name 4-hydroxy-5-(2-hydroxyacetamido)-2-{[3,4,5-trihydroxy-6-(4-methoxyphenoxy)oxan-2-yl]methoxy}-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
SMILES COC1=CC=C(OC2OC(COC3(CC(O)C(NC(=O)CO)C(O3)C(O)C(O)CO)C(O)=O)C(O)C(O)C2O)C=C1

Specifications

Color White
Quantity 5 mg
Formula Weight 593.54
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Neu5Gc alpha(2-6)Gal beta MP Glycoside
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.