accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1072946-52-1

IUPAC Name: (5-chloro-2-cyanophenyl)boronic acid
Synonyms: 5-chloro-2-cyanophenylboronic acid
Molecular Weight (g/mol): 181.38
Molecular Formula: C7H5BClNO2
InChi Key: JXIINQQPEHZQAY-UHFFFAOYSA-N
SMILES: OB(O)C1=C(C=CC(Cl)=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results