accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1072946-64-5

IUPAC Name: [5-cyano-2-(trifluoromethoxy)phenyl]boronic acid
Synonyms: 5-cyano-2-(trifluoromethoxy)phenylboronic acid
Molecular Weight (g/mol): 230.94
Molecular Formula: C8H5BF3NO3
InChi Key: SZPKBNMEIASWNB-UHFFFAOYSA-N
SMILES: OB(O)C1=C(OC(F)(F)F)C=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results