Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyano-2-(trifluoromethoxy)benzeneboronic acid, 97%, Thermo Scientific™

Catalog No. AAH61177MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61177MD 250 mg
AAH6117703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61177MD Supplier Thermo Scientific Chemicals Supplier No. H61177MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072946-64-5
Molecular Formula C8H5BF3NO3
Molecular Weight (g/mol) 230.937
InChI Key SZPKBNMEIASWNB-UHFFFAOYSA-N
Synonym 5-cyano-2-trifluoromethoxy phenylboronic acid, 5-cyano-2-trifluoromethoxy phenyl boronic acid, acmc-2098sk
PubChem CID 46738976
IUPAC Name [5-cyano-2-(trifluoromethoxy)phenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C#N)OC(F)(F)F)(O)O

Specifications

Quantity 250 mg
Formula Weight 230.94
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Cyano-2-(trifluoromethoxy)benzeneboronic acid
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.