accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOOO

CAS RN 1073-05-8

IUPAC Name: 1,3,2λ⁶-dioxathiane-2,2-dione
Synonyms: 1,3,2λ⁶-dioxathiane-2,2-dione
Molecular Weight (g/mol): 138.14
Molecular Formula: C3H6O4S
InChi Key: OQYOVYWFXHQYOP-UHFFFAOYSA-N
SMILES: O=S1(=O)OCCCO1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)