Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,2-Dioxathiane 2,2-Dioxide 98.0+%, TCI America™
SDP

Catalog No. D44275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D44275G Supplier TCI America Supplier No. D44275G
Add to Cart
Add to Cart

Specifications

CAS 1073-05-8
Melting Point 61°C
Color White
Boiling Point 153°C
Molecular Formula C3H6O4S
Quantity 5 g
MDL Number MFCD00801144
Synonym 1,3-Propylene Sulfate, 1,3-Propanediol Cyclic Sulfate, Trimethylene Sulfate
InChI Key OQYOVYWFXHQYOP-UHFFFAOYSA-N
SMILES C1COS(=O)(=O)OC1
IUPAC Name 1,3,2-dioxathiane 2,2-dioxide
Molecular Weight (g/mol) 138.137
PubChem CID 70623
Formula Weight 138.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,2-Dioxathiane 2,2-Dioxide
Show More Show Less
RTECSNumber TY4560000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.