accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH2

CAS RN 1073-67-2

IUPAC Name: 1-chloro-4-ethenylbenzene
Synonyms: P-chlorostyrene
Molecular Weight (g/mol): 138.59
Molecular Formula: C8H7Cl
InChi Key: KTZVZZJJVJQZHV-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C)C=C1
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)