Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Chlorostyrene

Catalog No. C7120310G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C7120310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C7120310G Supplier Sigma Aldrich Supplier No. C7120310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Chlorostyrene is a para-halogenated styrene derivative. It undergoes graft copolymerization with acrylonitrile (AN) onto ethylene-propylene-diene terpolymer (EPDM) in the presence of benzoyl peroxide (initiator).4-Chlorostyrene was used in the following studies:Study of chemical and biochemical properties of the vinyl group of styrene by the development of structure activity relationships (SAR). Preparation of new bis(pyrazolyl)borato olefin complexes of copper(I).To investigate the regioselectivity in the cationic Heck reaction of 4-substituted styrenes.

Specifications

CAS 1073-67-2
Density 1.155 g/mL (at 25°C)
Boiling Point 192°C
Molecular Formula C8H7Cl
Refractive Index n20/D 1.565 (literature)
Linear Formula H2C=CHC6H4Cl
MDL Number MFCD00000632
Quantity 10 g
Molecular Weight (g/mol) 138.59
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.