accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1073062-59-5

IUPAC Name: 2-(3,5-dibromophenyl)-4,6-diphenyl-1,3,5-triazine
Synonyms: 2-(3,5-dibromophenyl)-4,6-diphenyl-1,3,5-triazine
Molecular Weight (g/mol): 467.16
Molecular Formula: C21H13Br2N3
InChi Key: DRQMSTXYCLCAHO-UHFFFAOYSA-N
SMILES: BrC1=CC(=CC(Br)=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results