Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3,5-Dibromophenyl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. D4902200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4902200MG 200 mg
D49021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4902200MG Supplier TCI America Supplier No. D4902200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1073062-59-5
Molecular Formula C21H13Br2N3
Molecular Weight (g/mol) 467.164
MDL Number MFCD25562933
InChI Key DRQMSTXYCLCAHO-UHFFFAOYSA-N
PubChem CID 58382615
IUPAC Name 2-(3,5-dibromophenyl)-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC(=CC(=C3)Br)Br)C4=CC=CC=C4

Specifications

Melting Point 246°C
Color Yellow
Quantity 200 mg
Formula Weight 467.16
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3,5-Dibromophenyl)-4,6-diphenyl-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.