accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNN

CAS RN 1073354-41-2

IUPAC Name: 2-(pyrrolidin-1-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-(pyrrolidin-1-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 274.17
Molecular Formula: C15H23BN2O2
InChi Key: RKTXCUGUPDEZHR-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(N=CC=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results