CAS RN 1073354-73-0
IUPAC Name:
N-cyclohexyl-N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Synonyms:
N-cyclohexyl-N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular Weight (g/mol):
316.25
Molecular Formula:
C18H29BN2O2
InChi Key:
YTYXLMDYYHVPOF-UHFFFAOYSA-N
SMILES:
CN(C1CCCCC1)C1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
Sigma Aldrich 2-(Cyclohexylmethylamino)pyridine-5-boronic acid pinacol ester
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.