accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNNH2

CAS RN 1073354-99-0

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine
Molecular Weight (g/mol): 220.08
Molecular Formula: C11H17BN2O2
InChi Key: DAISWHFZWZZBBD-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(N)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results