accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNO

CAS RN 1073371-81-9

IUPAC Name: 2-(benzyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-(benzyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 311.19
Molecular Formula: C18H22BNO3
InChi Key: IFMSDMPTVPMGOP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(OCC2=CC=CC=C2)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results