accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3COOCH3OH

CAS RN 1073371-99-9

IUPAC Name: methyl 2-hydroxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Synonyms: methyl 2-hydroxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Molecular Weight (g/mol): 278.11
Molecular Formula: C14H19BO5
InChi Key: SRYGMLVCVMGUTB-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(O)C=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results