accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHGeH3HClClH

CAS RN 1074-29-9

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 262.13
Molecular Formula: C6H11Cl3Ge
InChi Key: CTVLIEBAMNPYNE-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.[GeH3]C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results