Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylgermanium Trichloride 98.0+%, TCI America™
SDP

Catalog No. P10031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P10031G Supplier TCI America Supplier No. P10031G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1074-29-9
Molecular Formula C6H5Cl3Ge
Molecular Weight (g/mol) 256.086
MDL Number MFCD00000460
InChI Key CIWQSBMDFPABPQ-UHFFFAOYSA-N
Synonym Phenyltrichlorogermane
PubChem CID 66176
IUPAC Name trichloro(phenyl)germane
SMILES C1=CC=C(C=C1)[Ge](Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 106°C
Quantity 1 g
UN Number 3265
Formula Weight 256.09
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Phenylgermanium Trichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.