accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OClO

CAS RN 107474-79-3

IUPAC Name: 1-(9H-fluoren-9-yl)ethyl carbonochloridate
Synonyms: 1-(9H-fluoren-9-yl)ethyl carbonochloridate
Molecular Weight (g/mol): 272.73
Molecular Formula: C16H13ClO2
InChi Key: SFRVOKMRHPQYGE-UHFFFAOYNA-N
SMILES: CC(OC(Cl)=O)C1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results