accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1075-38-3

IUPAC Name: 1-tert-butyl-3-methylbenzene
Synonyms: 1-tert-butyl-3-methylbenzene
Molecular Weight (g/mol): 148.25
Molecular Formula: C11H16
InChi Key: JTIAYWZZZOZUTK-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results