Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-tert-Butyltoluene, 99%, Thermo Scientific™

Catalog No. AAH3266806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3266806 5 g
AAH3266803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3266806 Supplier Thermo Scientific Chemicals Supplier No. H3266806

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075-38-3
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00059210
InChI Key JTIAYWZZZOZUTK-UHFFFAOYSA-N
Synonym 3-tert-butyltoluene, toluene, m-tert-butyl, benzene, 1-1,1-dimethylethyl-3-methyl, 1-tert-butyl-3-methylbenzene, m-tert-butyltoluene, 1-methyl-3-tert-butylbenzene, butyltoluol, acmc-1bo1t, 1-methyl-3-tert-butyl-benzene, jtiaywzzzozutk-uhfffaoysa
PubChem CID 33711
IUPAC Name 1-tert-butyl-3-methylbenzene
SMILES CC1=CC(=CC=C1)C(C)(C)C

Specifications

Melting Point -41°C
Boiling Point 189°C
Odor Aromatic
Quantity 5 g
UN Number UN2667
Formula Weight 148.25
Percent Purity 99%
Chemical Name or Material 3-tert-Butyltoluene
Hazard Category Danger
Hazard Statement GHS H Statement
H331-H227-H302-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BUTYLTOLUENES
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.