accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1075-62-3

IUPAC Name: N-(6-aminopyridin-2-yl)acetamide
Synonyms: N-(6-aminopyridin-2-yl)acetamide
Molecular Weight (g/mol): 151.17
Molecular Formula: C7H9N3O
InChi Key: CCVCJYWIEHEXOR-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(N)=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results