Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetamido-6-aminopyridine 98.0+%, TCI America™
SDP

Catalog No. A27075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27075G Supplier TCI America Supplier No. A27075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1075-62-3
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.17
MDL Number MFCD01099052
InChI Key CCVCJYWIEHEXOR-UHFFFAOYSA-N
PubChem CID 3767345
IUPAC Name N-(6-aminopyridin-2-yl)acetamide
SMILES CC(=O)NC1=CC=CC(N)=N1

Specifications

Melting Point 160°C
Color White
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-6-aminopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.