accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSN

CAS RN 107559-02-4

IUPAC Name: 2-methyl-6-phenyl-1,3-benzothiazole
Synonyms: 2-methyl-6-phenyl-1,3-benzothiazole
Molecular Weight (g/mol): 225.31
Molecular Formula: C14H11NS
InChi Key: QHLXZULIVMKONH-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=C(C=C2S1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results