Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-6-phenylbenzothiazole, 97%, Thermo Scientific™

Catalog No. AC329570010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC329570010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC329570010 Supplier Thermo Scientific Chemicals Supplier No. 329570010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 107559-02-4
Molecular Formula C14H11NS
Molecular Weight (g/mol) 225.31
MDL Number MFCD00086016
InChI Key QHLXZULIVMKONH-UHFFFAOYSA-N
Synonym 2-methyl-6-phenylbenzothiazole, 2-methyl-6-phenylbenzo d thiazole, benzothiazole, 2-methyl-6-phenyl, zlchem 705, acmc-20devf, pubchem15768, benzothiazole, 2-methyl-6-phenyl-6ci,9ci
PubChem CID 285329
IUPAC Name 2-methyl-6-phenyl-1,3-benzothiazole
SMILES CC1=NC2=CC=C(C=C2S1)C1=CC=CC=C1

Specifications

Melting Point 90°C to 95°C
Boiling Point 194.7°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 225.31
Percent Purity 97%
Chemical Name or Material 2-Methyl-6-phenylbenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.