accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONO

CAS RN 1076-59-1

IUPAC Name: 3-phenyl-4,5-dihydro-1,2-oxazol-5-one
Synonyms: 3-phenyl-4H-1,2-oxazol-5-one
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: IHKNLPPRTQQACK-UHFFFAOYSA-N
SMILES: O=C1CC(=NO1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results