accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSOOOFFFSOOOFFFSOOOFFFSOOOCe

CAS RN 107792-63-2

IUPAC Name: λ⁴-cerium(4+) tetratrifluoromethanesulfonate
Synonyms: λ⁴-cerium(4+) tetratriflate
Molecular Weight (g/mol): 736.37
Molecular Formula: C4CeF12O12S4
InChi Key: BYAJTISDHIASIP-UHFFFAOYSA-J
SMILES: [Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results